Learn More
4-Chloro-2,6-difluorobenzyl bromide, 97%, Thermo Scientific™
Product Code. 11392659
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392659 5 g 5g
11382659 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11392659 Supplier Thermo Scientific Alfa Aesar Supplier No. H26707.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Used frequently in organic synthesis. Find application as propellants.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 537013-52-8
Molecular Formula C7H4BrClF2
Molecular Weight (g/mol) 241.46
MDL Number MFCD03094463
InChI Key KYKUEAHGMDKVLF-UHFFFAOYSA-N
Synonym 2-bromomethyl-5-chloro-1,3-difluorobenzene, 4-chloro-2,6-difluorobenzyl bromide, 2-bromomethyl-5-chloro-1,3-difluoro-benzene, acmc-1b1j7, benzene, 2-bromomethyl-5-chloro-1,3-difluoro, 2-bromomethyl-5-chloranyl-1,3-bis fluoranyl benzene
PubChem CID 2773551
IUPAC Name 2-(bromomethyl)-5-chloro-1,3-difluorobenzene
SMILES FC1=CC(Cl)=CC(F)=C1CBr
Specifications
Quantity 5 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 241.46
Percent Purity 97%
Chemical Name or Material 4-Chloro-2,6-difluorobenzyl bromide