accessibility menu, dialog, popup

UserName

CAS RN 5381-24-8

SHO

CAS RN 5381-24-8

IUPAC Name: (1-benzothiophen-3-yl)methanol
Synonyms: 1-benzothiophen-3-ylmethanol
Molecular Weight (g/mol): 164.22
Molecular Formula: C9H8OS
InChi Key: UYGMKSKKGSUAHB-UHFFFAOYSA-N
SMILES: OCC1=CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results