accessibility menu, dialog, popup

UserName

HOCH

CAS RN 5390-04-5

IUPAC Name: pent-4-yn-1-ol
Synonyms: pent-4-yn-1-ol
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: CRWVOXFUXPYTRK-UHFFFAOYSA-N
SMILES: OCCCC#C
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)