accessibility menu, dialog, popup

UserName

CAS RN 5424-01-1

NH2NNHOO

CAS RN 5424-01-1

IUPAC Name: 3-aminopyrazine-2-carboxylic acid
Synonyms: 3-aminopyrazine-2-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C5H5N3O2
InChi Key: ZAGZIOYVEIDDJA-UHFFFAOYSA-N
SMILES: NC1=NC=CN=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results