accessibility menu, dialog, popup

UserName

CH3NCH3NOH(Z)

CAS RN 54376-65-7

IUPAC Name: N,N-diethyl-4-[(Z)-(hydroxyimino)methyl]aniline
Synonyms: N,N-diethyl-4-[(Z)-(hydroxyimino)methyl]aniline
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: DHNWBEYWLRLFHM-XFXZXTDPSA-N
SMILES: CCN(CC)C1=CC=C(\C=N/O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results