Learn More
4-Diethylaminobenzaldoxime, 97%, Thermo Scientific™
Product Code. 11342127
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342127 25 g 25g
11332127 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11342127 Supplier Thermo Scientific Alfa Aesar Supplier No. L07961.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54376-65-7
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00051801
InChI Key DHNWBEYWLRLFHM-FMIVXFBMSA-N
Synonym 4-diethylaminobenzaldoxime, 4-diethylamino benzaldehyde oxime, n,n-diethyl-4-1e-hydroxyimino methyl aniline, 4-diethylaminobenzaldehyde oxime, n,n-diethyl-4-hydroxyimino methyl aniline, 4-diethylaminobenzaldehydeoxime, diethyl 4-hydroxyimino methyl phenyl amine, n,n-diethyl-4-e-hydroxyimino methyl aniline, n,n-diethyl-4-1z-hydroxyimino methyl aniline, ne-n-4-diethylamino phenyl methylidene hydroxylamine
PubChem CID 6876690
IUPAC Name (NE)-N-[[4-(diethylamino)phenyl]methylidene]hydroxylamine
SMILES CCN(CC)C1=CC=C(C=C1)C=NO
Specifications
Melting Point 94°C to 97°C
Quantity 25 g
Beilstein 2722071
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material 4-Diethylaminobenzaldoxime