accessibility menu, dialog, popup

UserName

OHON

CAS RN 5462-71-5

IUPAC Name: 2-(4-cyanophenyl)acetic acid
Synonyms: (4-cyanophenyl)acetic acid
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: WEBXRQONNWEETE-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results