Learn More
4-Cyanophenylacetic acid, 97%
Product Code. 11371999
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371999 5 g 5g
11361999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11371999 Supplier Thermo Scientific Alfa Aesar Supplier No. H26062.06
Request Bulk or Custom Format
CAS: 5462-71-5 | C9H7NO2 | 161.16 g/mol
4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
Solubility
Soluble in CHCl3, THF, dioxane.
Notes
Storage temperature is 2 - 4°C and Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5462-71-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD06798066
InChI Key WEBXRQONNWEETE-UHFFFAOYSA-N
Synonym 2-4-cyanophenyl acetic acid, 4-cyanophenylacetic acid, 4-cyanophenyl acetic acid, benzeneacetic acid, 4-cyano, 4-cyano-phenyl-acetic acid, p-cyanophenylacetic acid, 4-cyanobenzeneacetic acid, p-cyanophenyl acetic acid, acetic acid, p-cyanophenyl
PubChem CID 79587
IUPAC Name 2-(4-cyanophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(C=C1)C#N
Specifications
Melting Point 150°C to 152°C
Quantity 5 g
UN Number UN3439
Solubility Information Soluble in CHCl3,THF,dioxane.
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 4-Cyanophenylacetic acid