accessibility menu, dialog, popup

UserName

CAS RN 54696-05-8

H3COO

CAS RN 54696-05-8

IUPAC Name: 1-[4-(benzyloxy)phenyl]ethan-1-one
Synonyms: 1-[4-(benzyloxy)phenyl]ethanone
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: MKYMYZJJFMPDOA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)