accessibility menu, dialog, popup

UserName

CAS RN 54745-92-5

ClONN

CAS RN 54745-92-5

IUPAC Name: quinoxaline-2-carbonyl chloride
Synonyms: quinoxaline-2-carbonyl chloride
Molecular Weight (g/mol): 192.6
Molecular Formula: C9H5ClN2O
InChi Key: SOPDQKNXOCUBSR-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CN=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results