accessibility menu, dialog, popup

UserName

H3CNHNOSOHNNSOCH3CH3

CAS RN 552309-42-9

IUPAC Name: 2-[(5-ethyl-6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)sulfanyl]-N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]acetamide
Synonyms: 2-[(5-ethyl-4-methyl-6-oxo-3H-pyrimidin-2-yl)sulfanyl]-N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]acetamide
Molecular Weight (g/mol): 416.51
Molecular Formula: C19H20N4O3S2
InChi Key: QSIYTNYMBWYHAA-UHFFFAOYSA-N
SMILES: CCC1=C(C)NC(SCC(=O)NC2=NC(=CS2)C2=CC=C(OC)C=C2)=NC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results