accessibility menu, dialog, popup

UserName

CH3NH3CON(E/Z)

CAS RN 55314-16-4

IUPAC Name: 3-(dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one
Synonyms: 3-(dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: MZLRFUCMBQWLNV-UHFFFAOYSA-N
SMILES: CN(C)C=CC(=O)C1=CN=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)