CAS RN 55699-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Oxymetazoline EP Impurity E, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-tert-Butyl-3-hydroxy-2,6-dimethylphenylacetonitrile, TRC
CAS: 55699-10-0 Molecular Formula: C14 H19 N O Molecular Weight (g/mol): 217.31 Synonym: [4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetonitrile,Oxymetazoline Hydrochloride Imp. E (EP) IUPAC Name: 2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetonitrile SMILES: Cc1cc(c(O)c(C)c1CC#N)C(C)(C)C