accessibility menu, dialog, popup

UserName

NOHF

CAS RN 55758-32-2

IUPAC Name: 6-fluoropyridin-3-ol
Synonyms: 6-fluoropyridin-3-ol
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: HTRLNWYWOKWCLV-UHFFFAOYSA-N
SMILES: OC1=CN=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results