accessibility menu, dialog, popup

UserName

ClSOOHNOHO

CAS RN 565198-64-3

IUPAC Name: 2-(3-chlorobenzenesulfonamido)acetic acid
Synonyms: (3-chlorobenzenesulfonamido)acetic acid
Molecular Weight (g/mol): 249.67
Molecular Formula: C8H8ClNO4S
InChi Key: VQWFYPPYALIWLV-UHFFFAOYSA-N
SMILES: OC(=O)CNS(=O)(=O)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results