CAS RN 5672-83-3
CAS RN 5672-83-3
IUPAC Name:
(4S)-4-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoate
Synonyms:
(4S)-4-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoate
Molecular Weight (g/mol):
294.28
Molecular Formula:
C14H16NO6
InChi Key:
BGMCTGARFXPQML-NSHDSACASA-M
SMILES:
COC(=O)[C@H](CCC([O-])=O)NC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester, 98%
CAS: 5672-83-3 Molecular Formula: C14H17NO6 Molecular Weight (g/mol): 295.291 MDL Number: MFCD00083278 InChI Key: BGMCTGARFXPQML-NSHDSACASA-N Synonym: z-glu-ome,z-l-glu-ome,1-methyl n-carbobenzoxy-l-glutamate,4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid,cbz-glu-ome,l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester,n-cbz-l-glu-ome,z-l-glutamic acid 1-methyl ester,n-cbz-l-glutamic acid 1-methyl ester,n-carbobenzoxy-l-glutamic acid 1-methyl ester PubChem CID: 2733408 IUPAC Name: (4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid SMILES: COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1