missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester, 98%

Product Code. 15450646
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15450646

Brand: Thermo Scientific Alfa Aesar H53417.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5672-83-3 | C14H17NO6 | 295.291 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5672-83-3
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.291
MDL Number MFCD00083278
InChI Key BGMCTGARFXPQML-NSHDSACASA-N
Synonym z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester
PubChem CID 2733408
IUPAC Name (4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 64°C to 70°C
Refractive Index 1.4365
Quantity 5 g
Beilstein 2567599
Optical Rotation −24° (c=1 in Ethanol)
Formula Weight 295.29
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.