CAS RN 56974-61-9
CAS RN 56974-61-9
IUPAC Name:
ethyl 4-[(6-{[amino(azaniumyl)methylidene]amino}hexanoyl)oxy]benzoate methanesulfonate
Synonyms:
ethyl 4-[(6-{[amino(ammonio)methylidene]amino}hexanoyl)oxy]benzoate mesylate
Molecular Weight (g/mol):
417.48
Molecular Formula:
C17H27N3O7S
InChi Key:
DNTNDFLIKUKKOC-UHFFFAOYSA-N
SMILES:
CS([O-])(=O)=O.CCOC(=O)C1=CC=C(OC(=O)CCCCCN=C(N)[NH3+])C=C1
1
–
3
of
3
results
1
Gabexate mesylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ Gabexate mesylate
CAS: 56974-61-9 Molecular Formula: C17H27N3O7S
3
Gabexate mesylate, MedChemExpress
MedChemExpress Gabexate mesylate is a Factor X inhibitor; serine protease inhibitor .