accessibility menu, dialog, popup

UserName

H3CCH3CH3OONHSCH3CH3CH3OHO

CAS RN 56976-06-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(tert-butylsulfanyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(tert-butylsulfanyl)propanoic acid
Molecular Weight (g/mol): 277.38
Molecular Formula: C12H23NO4S
InChi Key: OGARKMDCQCLMCS-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CSC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results