CAS RN 570-08-1
CAS RN 570-08-1
IUPAC Name:
1,3-diethyl 2-acetylpropanedioate
Synonyms:
1,3-diethyl 2-acetylpropanedioate
Molecular Weight (g/mol):
202.21
Molecular Formula:
C9H14O5
InChi Key:
SQAUUQRBOCJRCW-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(C(C)=O)C(=O)OCC
1
–
3
of
3
results
1
Diethyl 2-Acetylmalonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diethyl 2-Acetylmalonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Diethyl 2-Acetylmalonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.