CAS RN 575-36-0
IUPAC Name:
N-(naphthalen-1-yl)acetamide
Synonyms:
N-(naphthalen-1-yl)acetamide
Molecular Weight (g/mol):
185.23
Molecular Formula:
C12H11NO
InChi Key:
OKQIEBVRUGLWOR-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=C2C=CC=CC2=CC=C1
1
–
3
of
3
results
1
1-Acetamidonaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Acetamidonaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Acetamidonaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.