accessibility menu, dialog, popup

UserName

CAS RN 57710-57-3

CH3H3C(S)O(S)(S)OOHOO

CAS RN 57710-57-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results