missing translation for 'onlineSavingsMsg'
Learn More

Heptelidic Acid, MP Biomedicals™

Product Code. 15937305 Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.25 mg
1 mg
Unit Size:
1mg
250µg
This item is not returnable. View return policy

Product Code. 15937305

Brand: MP Biomedicals 0218337101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective, irreversible GAPDH inhibitor, as well as a DNA polymerase inhibitor. The koningic acid molecule, produced by fungi that consume sweet potatoes, has been shown to curb the excessive glucose consumption in tumors exhibiting the Warburg effect and leaving healthy cells alone.

  • Glyceraldehyde 3-phosphate dehydrogenase (GAPDH), a key enzyme in carbohydrate metabolism, reversibly catalyzes the conversion of GAP to 1,3-bisphosphoglycerate and NAD+
  • A sesquiterpene lactone produced by the fungus T. koningii that was shown to have antibiotic activity against anaerobic bacteria such as Bacteroides
  • It acts as an irreversible inhibitor of GAPDH that binds to the cysteine-149 residue at the active site of the enzyme (Ki = 1.6 μM)
  • Can selectively induce apoptosis in high-glycolytic cancer cells by inhibiting the generation of ATP in the glycolytic pathway
  • Heptelidic acid is also a selective and competitive inhibitor of mammalian DNA polymerases β and λ as well as terminal deoxynucleotidyl transferase in family X of DNA polymerases (Kis range from 5.2-9.5 μM)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57710-57-3
Molecular Formula C15H20O5
Synonym Koningic Acid

Specifications

Quantity 1 mg
Color White
Purity >95
Molecular Weight (g/mol) 280.32
Chemical Name or Material Heptelidic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.