CAS RN 57917-63-2
CAS RN 57917-63-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-[(1RS)-1-Ethyl-8-(1-methylethyl)-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Isopropyl Etodolac), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
8-Isopropyl Etodolac, TRC
CAS: 57917-63-2 Molecular Formula: C18 H23 N O3 Molecular Weight (g/mol): 301.38 Synonym: 2-[(1RS)-1-Ethyl-8-(1-methylethyl)-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid,Etodolac Imp. D (EP),8-Isopropyl Etodolac IUPAC Name: 2-(1-ethyl-8-propan-2-yl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid SMILES: CCC1(CC(=O)O)OCCc2c1[nH]c3c(cccc23)C(C)C