missing translation for 'onlineSavingsMsg'
Learn More

8-Isopropyl Etodolac, TRC

Product Code. 30406796
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406796 5 mg
30392494 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406796 Supplier Toronto Research Chemicals Supplier No. TRCI8245055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57917-63-2
Molecular Formula C18 H23 N O3
Molecular Weight (g/mol) 301.38
Synonym 2-[(1RS)-1-Ethyl-8-(1-methylethyl)-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, Etodolac Imp. D (EP), 8-Isopropyl Etodolac
IUPAC Name 2-(1-ethyl-8-propan-2-yl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid
SMILES CCC1(CC(=O)O)OCCc2c1[nH]c3c(cccc23)C(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H23NO3/c1-4-18(10-15(20)21)17-14(8-9-22-18)13-7-5-6-12(11(2)3)16(13)19-17/h5-7,11,19H,4,8-10H2,1-3H3,(H,20,21)
Formula Weight 301.1678
Chemical Name or Material 2-[(1RS)-1-Ethyl-8-(1-methylethyl)-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Isopropyl Etodolac)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.