accessibility menu, dialog, popup

UserName

CAS RN 586-98-1

HON

CAS RN 586-98-1

IUPAC Name: (pyridin-2-yl)methanol
Synonyms: 2-pyridinemethanol
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: SHNUBALDGXWUJI-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=N1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)