accessibility menu, dialog, popup

UserName

HClCH3H3CNO

CAS RN 58753-54-1

IUPAC Name: hydrogen 3-[benzyl(propan-2-yl)amino]-1-(naphthalen-2-yl)propan-1-one chloride
Synonyms: hydrogen 3-[benzyl(isopropyl)amino]-1-(naphthalen-2-yl)propan-1-one chloride
Molecular Weight (g/mol): 367.92
Molecular Formula: C23H26ClNO
InChi Key: NJTUORMLOPXPBY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)N(CCC(=O)C1=CC=C2C=CC=CC2=C1)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results