accessibility menu, dialog, popup

UserName

CAS RN 592-06-3

PtCNCN

CAS RN 592-06-3

IUPAC Name: platinum(2+) bis(iminomethanide)
Synonyms: platinum(2+) bis(cyanide)
Molecular Weight (g/mol): 247.12
Molecular Formula: C2N2Pt
InChi Key: INXLGDBFWGBBOC-UHFFFAOYSA-N
SMILES: [Pt++].[C-]#N.[C-]#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results