CAS RN 5958-24-7
CAS RN 5958-24-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Chlordiazepoxide EP Impurity B, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6-Chloro-2-(chloromethyl)-4-phenyl-quinazoline, TRC
CAS: 5958-24-7 Molecular Formula: C15 H10 Cl2 N2 O Molecular Weight (g/mol): 305.16 Synonym: 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide,Chlordiazepoxide Hydrochloride Imp. B (EP) IUPAC Name: 6-chloro-2-(chloromethyl)-3-oxido-4-phenylquinazolin-3-ium SMILES: [O-][n+]1c(CCl)nc2ccc(Cl)cc2c1c3ccccc3