CAS RN 597-67-1
CAS RN 597-67-1
IUPAC Name:
ethoxytriethylsilane
Synonyms:
ethoxytriethylsilane
Molecular Weight (g/mol):
160.33
Molecular Formula:
C8H20OSi
InChi Key:
DFJDZTPFNSXNAX-UHFFFAOYSA-N
SMILES:
CCO[Si](CC)(CC)CC
1
–
1
of
1
results
1
Ethoxytriethylsilane, 97%
CAS: 597-67-1 Molecular Formula: C8H20OSi Molecular Weight (g/mol): 160.332 MDL Number: MFCD00054718 InChI Key: DFJDZTPFNSXNAX-UHFFFAOYSA-N Synonym: triethylethoxysilane,silane, ethoxytriethyl,ethoxy triethyl silane,acmc-1al7t PubChem CID: 284135 IUPAC Name: ethoxy(triethyl)silane SMILES: CCO[Si](CC)(CC)CC