CAS RN 59729-37-2
CAS RN 59729-37-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fexinidazole, MedChemExpress
MedChemExpress Fexinidazole (HOE 239) is an orally active, potent nitroimidazole antitrypanosomal drug. Fexinidazole shows trypanocidal activity against T. brucei subspecies and strains with IC50s of 0.7-3.3 μM (0.2-0.9 μg/ml). Fexinidazol has the potential for human sleeping sickness (HAT) caused by infection with T. brucei.