accessibility menu, dialog, popup

UserName

CH3NHNFFFBr

CAS RN 60061-68-9

IUPAC Name: 4-bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 4-bromo-3-methyl-5-(trifluoromethyl)-2H-pyrazole
Molecular Weight (g/mol): 229
Molecular Formula: C5H4BrF3N2
InChi Key: PDSOUBXNWWZCNB-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(=NN1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results