accessibility menu, dialog, popup

UserName

CAS RN 60627-09-0

FCeFFF

CAS RN 60627-09-0

IUPAC Name: λ⁴-cerium(4+) hydrate tetrafluoride
Synonyms: λ⁴-cerium(4+) hydrate tetrafluoride
Molecular Weight (g/mol): 234.13
Molecular Formula: CeF4H2O
InChi Key: LBKPYHDQRHZOTL-UHFFFAOYSA-J
SMILES: O.[F-].[F-].[F-].[F-].[Ce+4]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results