accessibility menu, dialog, popup

UserName

CH3OCH3HN

CAS RN 61019-05-4

IUPAC Name: 5-methoxy-7-methyl-1H-indole
Synonyms: 5-methoxy-7-methyl-1H-indole
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: YGPVRHHGKCQSIL-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C2NC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results