accessibility menu, dialog, popup

UserName

HOONHOHO

CAS RN 612-42-0

IUPAC Name: 2-[(carboxymethyl)amino]benzoic acid
Synonyms: 2-[(carboxymethyl)amino]benzoic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: PJUXPMVQAZLJEX-UHFFFAOYSA-N
SMILES: OC(=O)CNC1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results