missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Carboxyphenyl)glycine, 97%, Thermo Scientific Chemicals

Product Code. 11332809
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332809 5 g 5g
11322809 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332809 Supplier Thermo Scientific Alfa Aesar Supplier No. H27701.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-42-0
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00020445
InChI Key PJUXPMVQAZLJEX-UHFFFAOYSA-N
Synonym n-2-carboxyphenyl glycine, 2-carboxymethyl amino benzoic acid, n-carboxymethyl anthranilic acid, phenylglycine-o-carboxylic acid, benzoic acid, 2-carboxymethyl amino, 2-carboxymethyl-amino-benzoic acid, glycine, n-o-carboxyphenyl, 2-carboxymethylamino benzoic acid, anthranilic acid, n-carboxymethyl
PubChem CID 69161
IUPAC Name 2-(carboxymethylamino)benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NCC(=O)O

Specifications

Boiling Point ∼218°C (decomposition)
Quantity 5 g
Formula Weight 195.17
Percent Purity 97%
Chemical Name or Material N-(2-Carboxyphenyl)glycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.