CAS RN 612-61-3
IUPAC Name:
7-chloroquinoline
Synonyms:
7-chloroquinoline
Molecular Weight (g/mol):
163.6
Molecular Formula:
C9H6ClN
InChi Key:
QNGUPQRODVPRDC-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2C=CC=NC2=C1
1
–
2
of
2
results
1
7-Chloroquinoline, TRC
CAS: 612-61-3 Molecular Formula: C9 H6 Cl N Molecular Weight (g/mol): 163.6 IUPAC Name: 7-chloroquinoline SMILES: Clc1ccc2cccnc2c1
2
7-Chloroquinoline, 97%, Thermo Scientific™
CAS: 612-61-3 Molecular Formula: C9H6ClN Molecular Weight (g/mol): 163.61 InChI Key: QNGUPQRODVPRDC-UHFFFAOYSA-N Synonym: quinoline, 7-chloro,7-chloro-quinoline,pubchem19899 PubChem CID: 521963 IUPAC Name: 7-chloroquinoline SMILES: C1=CC2=C(C=C(C=C2)Cl)N=C1