missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroquinoline, TRC

Product Code. 30136455
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136455 1 g 1g
30011855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136455 Supplier Toronto Research Chemicals Supplier No. TRCC3813401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-61-3
Molecular Formula C9 H6 Cl N
Molecular Weight (g/mol) 163.6
IUPAC Name 7-chloroquinoline
SMILES Clc1ccc2cccnc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H6ClN/c10-8-4-3-7-2-1-5-11-9(7)6-8/h1-6H
Formula Weight 163.0189
Chemical Name or Material 7-Chloroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.