accessibility menu, dialog, popup

UserName

CAS RN 613-84-3

IUPAC Name: 2-hydroxy-5-methylbenzaldehyde
Synonyms: 2-hydroxy-5-methylbenzaldehyde
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: ILEIUTCVWLYZOM-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(C=O)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results