accessibility menu, dialog, popup

UserName

CAS RN 6132-37-2

H3COOOBr

CAS RN 6132-37-2

IUPAC Name: ethyl 5-bromofuran-2-carboxylate
Synonyms: ethyl 5-bromofuran-2-carboxylate
Molecular Weight (g/mol): 219.03
Molecular Formula: C7H7BrO3
InChi Key: GVAPLPBPJARJEY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Br)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results