missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromo-2-furoate, 97+%, Thermo Scientific™

Product Code. 15498997
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15498997

Brand: Thermo Scientific Alfa Aesar H52343.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-37-2
Molecular Formula C7H7BrO3
Molecular Weight (g/mol) 219.034
MDL Number MFCD00092362
InChI Key GVAPLPBPJARJEY-UHFFFAOYSA-N
Synonym ethyl 5-bromo-2-furoate, ethyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid ethyl ester, acmc-209mrg, ethyl 5-bromo-2-furate, 2-bromo-5-ethoxycarbonyl-furan, ethyl5-bromo-2-furoate,97+%, ethyl-5-bromofuran-2-carboxylate, ethyl 5-bromofuran-2-carboxylate, 97+%, 5-bromofuran-2-carboxylic acid ethyl ester
PubChem CID 345282
IUPAC Name ethyl 5-bromofuran-2-carboxylate
SMILES CCOC(=O)C1=CC=C(O1)Br

Specifications

Quantity 5 g
Formula Weight 219.03
Percent Purity ≥97%
Chemical Name or Material Ethyl 5-bromo-2-furoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.