CAS RN 61337-88-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
1-(3-Cyano-2-pyridyl)-4-methyl-2-phenylpiperazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(3-Cyano-2-pyridyl)-4-methyl-2-phenylpiperazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(3-Cyano-2-pyridyl)-4-methyl-2-phenylpiperazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-carbonitrile, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.