CAS RN 62148-67-8
CAS RN 62148-67-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Chlorobenzyl 4-Fluorophenyl Ketone, TRC
CAS: 62148-67-8 Molecular Formula: C14H10ClFO Molecular Weight (g/mol): 248.68 Synonym: 2-Chloro-1-(4-fluorophenyl)-2-phenylethanone IUPAC Name: 2-chloro-1-(4-fluorophenyl)-2-phenylethanone SMILES: Fc1ccc(cc1)C(=O)C(Cl)c2ccccc2