CAS RN 625-37-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-2-Butenamide, TRC
CAS: 625-37-6 Molecular Formula: C4 H7 N O Molecular Weight (g/mol): 85.1 IUPAC Name: (E)-but-2-enamide SMILES: C\C=C\C(=O)N