missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Butenamide, TRC

Product Code. 30240739
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240739 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30240739 Supplier Toronto Research Chemicals Supplier No. TRCB690200250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-37-6
Molecular Formula C4 H7 N O
Molecular Weight (g/mol) 85.1
IUPAC Name (E)-but-2-enamide
SMILES C\C=C\C(=O)N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H7NO/c1-2-3-4(5)6/h2-3H,1H3,(H2,5,6)/b3-2+
Formula Weight 85.0528
Chemical Name or Material trans-2-Butenamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.