CAS RN 627-08-7
CAS RN 627-08-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isopropyl Propyl Ether, TRC
CAS: 627-08-7 Molecular Formula: C6 H14 O Molecular Weight (g/mol): 102.175 Synonym: 1-(1-Methylethoxy)propane,1-(Propan-2-yloxy)propane,Isopropyl propyl ether,Propyl iso-propyl ether,Propyl isopropyl ether; IUPAC Name: 1-propan-2-yloxypropane SMILES: CCCOC(C)C