accessibility menu, dialog, popup

UserName

CAS RN 627-95-2

HClH2NOHO

CAS RN 627-95-2

IUPAC Name: hydrogen 5-aminopentanoic acid chloride
Synonyms: hydrogen 5-aminovaleric acid chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: BLOIUFYKQCCAGP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results