CAS RN 62929-69-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-O-(E)-Feruloylquinic Acid, TRC
CAS: 62929-69-5 Molecular Formula: C17H20O9 Molecular Weight (g/mol): 368.34 Synonym: [1S-[1alpha,3alpha,4alpha,5beta(E)]]-1,3,4-Trihydroxy-5-[[3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propenyl]oxy]cyclohexanecarboxylic Acid; IUPAC Name: (1S,3R,4R,5R)-1,3,4-Trihydroxy-5-(((E)-3-(4-hydroxy-3-methoxyphenyl)acryloyl)oxy)cyclohexane-1-carboxylic Acid SMILES: COc1cc(\C=C\C(=O)O[C@@H]2C[C@@](O)(C[C@@H](O)[C@H]2O)C(=O)O)ccc1O