accessibility menu, dialog, popup

UserName

CH3SOOOOSOOCH3

CAS RN 6315-52-2

IUPAC Name: 2-[(4-methylbenzenesulfonyl)oxy]ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-[(4-methylbenzenesulfonyl)oxy]ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 370.43
Molecular Formula: C16H18O6S2
InChi Key: LZIPBJBQQPZLOR-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCOS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)