accessibility menu, dialog, popup

UserName

CAS RN 638-39-1

SnCH3OOCH3OO

CAS RN 638-39-1

IUPAC Name: λ²-tin(2+) diacetate
Synonyms: λ²-tin(2+) diacetate
Molecular Weight (g/mol): 236.8
Molecular Formula: C4H6O4Sn
InChi Key: PNOXNTGLSKTMQO-UHFFFAOYSA-L
SMILES: [Sn++].CC([O-])=O.CC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results